Learn More
Perkin Elmer US LLC Double Aperature Accessory
Supplier:  Perkin Elmer US LLC L6310202
Encompass_Preferred
The photometric accuracy of spectrophotometers is limited by detector non-linearity errors. These errors can be reduced by use of the double aperture accessory, which allows non-linearity to be characterized and corrected for. Typical applications include specular reflectance measurements on very high reflectance laser mirrors and transmittance measurements on filters. Includes a manual containing step-by-step measurement procedures and full details of all calibration calculations.
Catalog No. 50-201-6507
Add to Cart
Request a Quote