Learn More
Perkin Elmer US LLC Fast Filter Accessory Installation Kit
Supplier:  Perkin Elmer US LLC L2250019
Encompass_Preferred
The plug-n-play Fast Filter accessory enables analysis of rapid intracellular ion movements into and out of cells (signal transduction) using ratiometric fluorescent probes. This accessory rapidly rotates a paired set of bandpass filters (not included) aligned to the fluorophore under investigation.
Catalog No. 50-201-6441