Learn More
Bachem Ac-Ala-Leu-Cys-Asp-Asp-Pro-Arg-Val-Asp-Arg-Trp-Tyr-Cys-Gln-Phe-Val-Glu-Gly-NH2
SDP
Supplier:  Bachem 4033397.0001
Encompass_Preferred
Ac-Ala-Leu-Cys-Asp-Asp-Pro-Arg-Val-Asp-Arg-Trp-Tyr-Cys-Gln-Phe-Val-Glu-Gly-NH2, 1926163-13-4, C97H139N27O29S2, Mr 2211.47, 1mg. E-76. The potent anticoagulant E-76, Acetyl-ALCDDPRVDRWYCQFVEG-amide, is an inhibitor of coagulation factor VIIa. It has been suggested that E-76 works by binding to an exosite on the factor VIIa protease domain, and non-competitively inhibits activation of factor X and amidolytic activity. E-76 has a well defined structure in solution determined by NMR spectroscopy that is similar to the X-ray crystal structure when complexed with factor VIIa and represents a new framework for the development of inhibitors for serine proteases.
Catalog No. 50-194-6750