Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ The Properties of Polymers
GREENER_CHOICESCI_ED

Catalog No. S96898 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96898 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96898 Supplier Innovating Science™ Supplier No. IS2505
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 4g Sodium Alginate (Alginic Acid); 30mL Calcium Chloride concentrate; 250mL 3% Polyvinyl Alcohol (4); 200mL 4% Sodium Borate; 35g Sodium Polyacrylate powder; clear plastic cups (45); 30mL graduated measuring cups (15); 15mL plastic pipets (15)

Requires

Electronic balance, graduated cylinders, stirring rods, distilled or deionized water, and food coloring (optional)

Investigate how polymers affect every aspect of daily life, from plastics to clothing.

  • Perform a cross-linking procedure on polymers to create polymer “worms” and the classic “slime”
  • Examine properties of a super-absorbent polymer capable of absorbing hundreds of times its own weight in water to create “snow”

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To examine properties of a super-absorbent polymer capable of absorbing hundreds of times its own weight in water to create snow
Includes 1 x 4g Sodium alginate (Alginic acid), 1 x 30mL Calcium Chloride concentrate, 4 x 250mL 3% Polyvinyl Alcohol, 1 x 200mL 4% Sodium Borate, 1 x 35g Sodium Polyacrylate Powder (snow), 45 clear Plastic cups, 15 30mL graduated measuring cups, 15 1mL Plastic pipettes, teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 12.125 x 9.25 x 6.5 in.
Class Size 15 Lab Groups
Product Type Properties of Polymers Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.