Learn More
Bachem DL-b-Phenyllactic acid
SDP
Catalog No. 501947206
Encompass_Preferred
DL-b-Phenyllactic acid, 828-01-3, C9H10O3, Mr 166.18, 25g. (RS)-2-Hydroxy-3-phenyl-propionic acid. The racemate and both enantiomers of PLA show antifungal activity.
Catalog No. 50-194-7206 Supplier Bachem Supplier No. 4000118.0025
DL-b-Phenyllactic acid, 828-01-3, C9H10O3, Mr 166.18, 25g. (RS)-2-Hydroxy-3-phenyl-propionic acid. The racemate and both enantiomers of PLA show antifungal activity.