Learn More
Braintree Scientific The Radial Arm-Maze, 8-Arm Maze, Mouse
SDP
Catalog No. 501954141
Encompass
The Radial Arm-Maze has been used extensively in evaluating spatial learning and memory. The apparatus consists of eight (or 12) equidistantly-spaced arms radiating from a small circular central platform. The design ensures that after checking for food at the end of each arm the rat is forced to return to the central platform before making another choice. The rat always has eight (or 12) possible options. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4141 Supplier Braintree Scientific Supplier No. 60150M
The Radial Arm-Maze has been used extensively in evaluating spatial learning and memory. The apparatus consists of eight (or 12) equidistantly-spaced arms radiating from a small circular central platform. The design ensures that after checking for food at the end of each arm the rat is forced to return to the central platform before making another choice. The rat always has eight (or 12) possible options. **Sold for research purposes only and not for human use**