Learn More
Perkin Elmer US LLC Rotating Sample Holder for Frontier External Mounted NIRA
Catalog No. 502016505
Encompass_Preferred
The Rotating Sample Holder for the Sample Compartment NIRA is designed to enhance reproducibility of measurements by enabling collection of a representative spectrum of an inhomogenous sample.
Catalog No. 50-201-6505 Supplier Perkin Elmer US LLC Supplier No. L1280317
The Rotating Sample Holder for the Sample Compartment NIRA is designed to enhance reproducibility of measurements by enabling collection of a representative spectrum of an inhomogenous sample.