Learn More
Perkin Elmer US LLC Rotating Sample Holder for Frontier Sample Compartment NIRA
Catalog No. 502016506
Encompass_Preferred
The Rotating Sample Holder for the Sample Compartment NIRA is designed to enhance reproducibility of measurements by enabling collection of a representative spectrum of an inhomogenous sample.
Catalog No. 50-201-6506 Supplier Perkin Elmer US LLC Supplier No. L1280314
The Rotating Sample Holder for the Sample Compartment NIRA is designed to enhance reproducibility of measurements by enabling collection of a representative spectrum of an inhomogenous sample.