Learn More
Enterprise Technology Solutions The Safety Zone Gloves,Flock-lined,Latex,Medium,18 mil,12"L,YW
SDP
Catalog No. 5000827794
Encompass_Preferred
Flock-lined Latex Gloves with a pinked cuff may be universally known as a dishwashing glove, but the uses extend beyond household cleaning. High-volume foodservice (such as meat-processing plants) benefits from an increased resistance to animal fats. The 18 mil thick gloves comply with Federal Food, Drug and Cosmetic Act provisions that allow repeated use in direct contact with food. For a better grip, the glove surface features a fish-scale texture. Gloves are 100 percent PFAS-free and 100 percent phthalate-free.
Catalog No. 50-008-27794 Supplier Enterprise Technology Solutions Supplier No. SZNGRFYMD1S
Flock-lined Latex Gloves with a pinked cuff may be universally known as a dishwashing glove, but the uses extend beyond household cleaning. High-volume foodservice (such as meat-processing plants) benefits from an increased resistance to animal fats. The 18 mil thick gloves comply with Federal Food, Drug and Cosmetic Act provisions that allow repeated use in direct contact with food. For a better grip, the glove surface features a fish-scale texture. Gloves are 100 percent PFAS-free and 100 percent phthalate-free.