Learn More
Enterprise Technology Solutions The Safety Zone Gloves,High-density Poly,Latex-free,Small,100/PK,CL
SDP
Supplier:  Enterprise Technology Solutions SZNGDPESM
Encompass_Preferred
High-density polyethylene gloves feature an embossed grip for better hold. They are most often used for light-duty tasks that require frequent glove changes. Powder-free, 11-3/4" long gloves have a loose fit for easy on-and-off applications and are especially useful in foodservice lines, deli counters and other high-volume applications. They comply with the provisions of the Federal Food, Drug and Cosmetic Act that allows repeated use in direct contact with food. Clear construction features die-cut and heat-sealed edges for dependable use. Gloves feature a 1 mil cuff thickness, 1 mil palm thickness and 1 mil finger thickness for more durable use.
Catalog No. 50-008-27895