Learn More
Enterprise Technology Solutions The Safety Zone Gloves, Medical-Grade Vinyl, PF, Small, 9-1/4"L,100/BX, WE
SDP
Supplier:  Enterprise Technology Solutions SZNGVEPSM1C
Encompass_Preferred
Premium, medical-grade, vinyl exam gloves are stronger than polyethylene gloves and offer greater comfort for jobs like medical applications and cleaning where gloves will be worn for a greater time period. They meet FDA 510K Standards for examination gloves used in medical applications and comply with Federal Food, Drug and Cosmetic Act provisions that allow repeated use in direct contact with food. Powder-free gloves go through an extra process of chlorinating the gloves to ease donning. They are also latex-free, DINP-free and DEHP-free for safer use. The 9-1/4" long gloves feature a 2.6 mil cuff thickness, 3.6 mil palm thickness and 4.1 mil finger thickness for more durable use.
Catalog No. 50-008-27780