Learn More
Enterprise Technology Solutions The Safety Zone Gloves,Nitrile,Flock-lined,13"L,Med,1 Pair/Bag,12 Bags/DZ,GN
SDP
Catalog No. 5000827807
Encompass_Preferred
These Flock-lined Nitrile Gloves may be universally known as "that dishwashing glove," but the uses extend beyond household cleaning. High-volume foodservice such as meat-processing plants benefit from their increased resistance to animal fats. Raised diamond grip on these 13" long gloves helps prevent slipping. Palm thickness is 15 mil. Long, straight cuff protects your more than your hands.
Catalog No. 50-008-27807 Supplier Enterprise Technology Solutions Supplier No. SZNGNGFMD15C
These Flock-lined Nitrile Gloves may be universally known as "that dishwashing glove," but the uses extend beyond household cleaning. High-volume foodservice such as meat-processing plants benefit from their increased resistance to animal fats. Raised diamond grip on these 13" long gloves helps prevent slipping. Palm thickness is 15 mil. Long, straight cuff protects your more than your hands.