Learn More
Enterprise Technology Solutions The Safety Zone Gloves, Polyurethane-Coated, 2X-Large, 12 PR/DZ, Gray
SDP
Catalog No. 5000827808
Encompass_Preferred
Knit gloves feature polyurethane-coated palms and fingers for increased strength and protection. Flexible material provides a comfortable fit for increased dexterity. Breathable design allows air to flow freely to the back of your hands to help keep hands cool during tasks. 2X-large gloves are perfect for a wide variety of industrial work applications.
Catalog No. 50-008-27808 Supplier Enterprise Technology Solutions Supplier No. SZNGNPU2X4GY
Knit gloves feature polyurethane-coated palms and fingers for increased strength and protection. Flexible material provides a comfortable fit for increased dexterity. Breathable design allows air to flow freely to the back of your hands to help keep hands cool during tasks. 2X-large gloves are perfect for a wide variety of industrial work applications.