Learn More
Enterprise Technology Solutions The Safety Zone Gloves, Powder-free, Vinyl, 9-1/4"L, Large, 100/BX, Clear
SDP
Catalog No. 5000827773
Encompass_Preferred
General-purpose vinyl gloves are stronger than polyethylene gloves and offer greater comfort for jobs like food preparation, bussing and cleaning where gloves will be worn for a greater period of time. All components comply with the provisions of the Federal Food, Drug and Cosmetic Act so they are safe for repeated use in direct contact with food. These economy gloves are produced with 100 percent vinyl resin so they are 100 percent latex-free. They are also 100 percent DEHP-free and DINP-free. Powder-free gloves go through an extra process of chlorinating the gloves to ease donning. Size large gloves measure about 9-1/4" long and offer a 3.5 mil finger thickness, 3.2 mil palm thickness and 2.0 mil cuff thickness.
Catalog No. 50-008-27773 Supplier Enterprise Technology Solutions Supplier No. SZNGVP9LGHH
General-purpose vinyl gloves are stronger than polyethylene gloves and offer greater comfort for jobs like food preparation, bussing and cleaning where gloves will be worn for a greater period of time. All components comply with the provisions of the Federal Food, Drug and Cosmetic Act so they are safe for repeated use in direct contact with food. These economy gloves are produced with 100 percent vinyl resin so they are 100 percent latex-free. They are also 100 percent DEHP-free and DINP-free. Powder-free gloves go through an extra process of chlorinating the gloves to ease donning. Size large gloves measure about 9-1/4" long and offer a 3.5 mil finger thickness, 3.2 mil palm thickness and 2.0 mil cuff thickness.