Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ The Science Behind Finding a Perfect Match : Blood Typing for Stem Cell Transplants
SDPSCI_ED

Catalog No. S001099 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S001099 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001099 Supplier EDVOTEK™ Supplier No. 142
Add to Cart
Add to Cart

Students will assume the role of transplant physicians by analyzing simulated blood samples for transplant compatibility and comparing genetic markers to identify which donor is the best match for a leukemia patient.

  • Students will analyze simulated blood samples using real-world techniques utilized in biomedical labs.
  • Learn to count white blood cells using a hemocytometer.
  • Perform blood compatibility tests to reveal information about donor blood samples.
  • Determine who is the best candidate to donate stem cells to a patient in need.
  • Group Size: For 10 Lab Groups.
  • Kit Includes: Instructions, Patient and Donor Simulated Blood Samples, Simulated White Blood Cell Counting Solutions, Anti-A Serum, Anti-B Serum, Anti-Rh Serum, AHG Serum, Red Dye Concentrate, Transfer Pipets, Microtiter Plates, Microcentrifuge Tubes, Counting Chambers.
  • All You Need: Micropipettes: 5-50 μL and tips, Microscopes, Distilled or Deionized Water, Bleach Solution or Lab Disinfectant.
  • Storage: Room Temperature.

Specifications

Grade Level High School, Higher Ed
Grade 9+
Class Size 10 Lab Groups
Product Type ABO-Rh Blood Typing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.