Learn More
Bachem Gluten Exorphin A5
SDP
Supplier:  Bachem 4025054.0025
Encompass_Preferred
Gluten Exorphin A5, H-Gly-Tyr-Tyr-Pro-Thr-OH, 142155-24-6, C29H37N5O9, Mr 599.64, 25mg. The sequence of this opioid peptide was found at 15 sites in the primary structure of the high molecular weight glutenin. Gluten exorphin A5 is highly specific for d-receptors and displays opioid activity in the MVD assay. The N-terminal Gly increases its activity - thus the structure-activity relationships of gluten exorphins A are quite different from those of the endogenous opioid peptides.
Catalog No. 50-194-6974