Learn More
Bachem Neurotensin (8-13) Acetate
SDP
Supplier:  Bachem 4100575.0025
Encompass_Preferred
Neurotensin (8-13) Acetate, H-Arg-Arg-Pro-Tyr-Ile-Leu-OH, 60482-95-3, C38H64N12O8, Mr 817.00, 25mg. The sequence RRPYIL is essential for neurotensin-NT receptor interaction.
Catalog No. 50-194-6984