Learn More
Bachem PHF6 Trifluoroacetate
SDP
Supplier:  Bachem 4062344.0500
Encompass_Preferred
PHF6 Trifluoroacetate, H-Val-Gln-Ile-Val-Tyr-Lys-OH, 329897-62-3, C36H60N8O9, Mr 748.92, 0.5mg. Paired helical filament-6 Tau Protein (306-311) (human). The sequence VQIVYK within the third repeat of tau is essential for fibrillization.
Catalog No. 50-194-6630