Learn More
Perkin Elmer US LLC Small-Spot Directional IV Absolute Reflectance Accessory
Catalog No. 502016501
Encompass_Preferred
The small-spot directional IV accessory is a modified version that has optics which are specially configured to measure small samples. Using this accessory, samples as small as 3 mm in height and 1 mm in width can be measured.
Catalog No. 50-201-6501 Supplier Perkin Elmer US LLC Supplier No. L6310208
Add to Cart
Add to Cart
Request a Quote
The small-spot directional IV accessory is a modified version that has optics which are specially configured to measure small samples. Using this accessory, samples as small as 3 mm in height and 1 mm in width can be measured.