Learn More
SOMNI Scientific™ The SOMNI Popoff Occlusion Valve
SDP
Catalog No. 04777179 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-179 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-179 Supplier SOMNI Scientific™ Supplier No. PK4015
Add to Cart
Add to Cart
Simple device every rebreathing or bain system needs. The occlusion valve allows for safe mechanical breath delivery without closing the popoff valve on the anesthesia machine. One handed operation
  • Simple device every rebreathing or bain system needs
  • Safe mechanical breath delivery without closing the popoff valve
  • One handed operation
  • Easy installation
  • 19 mm Female x 19 mm Male
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) Rebreathing or Bain System