Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company The Tank Oil Draining System
SCI_ED

Catalog No. S2337152 Shop All Fischer Technical Company Products
Change view
Click to view available options
For Use With (Equipment):
Works with Fischer Technical LAV Series Pumps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S2337152 Works with Fischer Technical LAV Series Pumps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2337152 Supplier Fischer Technical Company Supplier No. LAVTANK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Tank Oil Draining System. Holds up to 5 oil changes.

  • All Vacuum Pumps require changing oil after every use.
  • Contaminated oil, laden with moisture and acid will damage pumps and shorten pump life.
  • Deep vacuums cannot be achieved with contaminated oil.
  • Maintaining clean oil in your pump will ensure that your pump will operate at peak efficiency.
  • No more mess & spills environmentally friendly!
  • The Tank is easy to use: Turn black cap counterclockwise to open. Place Vacuum Pump in the cradle and drain pump. Once the pump has finished draining, turn the black cap clockwise to close after every use.
  • Holds up to 5 oil changes

Specifications

Color Black
For Use With (Application) Holding Multiple Oil Changes
Type Tank
Description Holds up to 5 Oil Changes
Material Plastic
For Use With (Equipment) Works with Fischer Technical LAV Series Pumps
No. per Case 1
No. per Pack 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.