Learn More
Perkin Elmer US LLC Anti-Convection Extension Tube - Qty. 1
Supplier:  Perkin Elmer US LLC N5191324
Encompass_Preferred
This extension tube connects to the Anti-Convection Tube in the TGA 7 High Temperature and the PYRIS 1 TGA High Temperature furnaces.
Catalog No. 50-201-6531