Learn More
Bachem H-Gly-Gly-Tyr-OH
SDP
Supplier:  Bachem 4007920.0005
Encompass_Preferred
H-Gly-Gly-Tyr-OH, 17343-07-6, C13H17N3O5, Mr 295.30, 5g. The tripeptide GGY showed ACE-inhibitory activity, IC50 1.3 umol/l.
Catalog No. 50-194-6752