Learn More
Electron Microscopy Sciences The Tweezer Repair Kit
SDP
Catalog No. 501927087
Encompass_Preferred
This repair kit allows you to straighten, resharpen, and repair your tweezers that have been damaged or have become dull. Instructions are included with the kit. The kit includes 1 sharpening stone, ten O-rings, instrument Oil, 50 ml, and chain nose pliers.
Catalog No. 50-192-7087 Supplier Electron Microscopy Sciences Supplier No. 7289210
This repair kit allows you to straighten, resharpen, and repair your tweezers that have been damaged or have become dull. Instructions are included with the kit. The kit includes 1 sharpening stone, ten O-rings, instrument Oil, 50 ml, and chain nose pliers.