Learn More
Anton Paar The Ultratap 500 series of tap density testers delivers unmatched durability with guaranteed 25 million taps and a three-year warranty for long-lasting reliability.They are also 90 % quieter, offering the lowest noise levels on the market.
Supplier:  Anton Paar 323323
Encompass_Preferred
Stations: 2Cylinder sizes accommodated: 250 mL standard,5, 10, 25, 50, 100, 250, 500, 1,000 mLTap speed: User configurable 50 to 350 taps/minuteDrop height: 3 mm (0.12 inch) ± 0.2 mm or 14 mm (0.55 inch) ± 1 mmSound level: 65 dB (58 dB with optional noise reduction cabinet)Tap mode: User configurable by time (1 second to 999 minutes) or number of taps (1 to 99,999)User interface: 7-inch TFT WVGA (800 x 480 pixels); PCAP touchscreenUser managementIncluded (with password protection)USB ports; 2Supported input devicesBarcode reader, keyboard, mouse, touchscreen, Balance and printer connection via USB portBuilt-in calculations: Bulk density, Carr’s compressibility index, Hausner ratio, tapped densityStorage capacity: 1,000 reports/100 methodsBuilt-in methods: 25
Catalog No. 50-005-80462