Learn More
Bachem (4-((2-Methylphenyl)aminocarbonyl)-aminophenyl)acetyl-Fibronectin CS-1 Fragment (1980-1983)
SDP
Supplier:  Bachem 4029156.0025
Encompass_Preferred
(4-((2-Methylphenyl)aminocarbonyl)-aminophenyl)acetyl-Fibronectin CS-1 Fragment (1980-1983), (4-((2-Methylphenyl)aminocarbonyl)-aminophenyl)acetyl-Leu-Asp-Val-Pro-OH, 187735-94-0, C36H48N6O9, Mr 708.81, 25mg. The VLA-4 inhibitor BIO-1211 based on the LDVP motif represents a selective, tight-binding inhibitor of integrin a4b1 achieved by appending an optimal organic group to the N-terminus of the peptide (Kd = 70 pM). Moreover, this inhibitor exhibited > 200-fold selectivity for the activated form of the integrin a4b1 receptor.
Catalog No. 50-194-6558