Learn More
Bachem 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOH
SDP
Supplier:  Bachem 4025806.0025
Encompass_Preferred
4-Abz-Gly-Pro-D-Leu-D-Ala-NHOH Trifluoroacetate, 124168-73-6, C23H34N6O6, Mr 490.56, 25mg. FN-439, MMP Inhibitor 1. The water-soluble peptidylhydroxamic acid FN439 is a broad-spectrum inhibitor of human matrix metalloproteinases. 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOH retains its activity even after prolonged incubation with pronase or human granulocyte elastase. FN 439 inhibits interstitial and granulocyte collagenases, granulocyte gelatinase and skin fibroblast stromelysin with IC50 of 1 · 10-6 M, 3 · 10-5 M and 1.5 · 10-4 M, respectively.
Catalog No. 50-194-6719