Learn More
Research Products International Corp Well Orienter System, Includes Orienter Card, Stand and Grid Pad
SDP
Supplier:  Research Products International Corp 248803
Encompass
The Well Orienter System increases accuracy and efficiency with 96 well micro-plate procedures. The complete system contains a well index card, plate stand and pre-printed pad simulating a 96 well grid layout. The plastic well index card fits perfectly under a micro-plate and clearly identifies each well with distinct alpha/numeric characters. The plate stand holds an index card and micro-plate at the optimum viewing angle to view for pipetting while sitting or standing. Stand has rubber feet and a neoprene backing to prevent slipping.The pre-printed 96 well grid pad allows convenient recording or sample data. Each grid has a mild adhesive strip for affixing to a laboratory notebook page.
Catalog No. 50-550-314