Learn More
TARGETMOL CHEMICALS INC THEAFLAVIN-3-GALLATE 5MG
SDP
Catalog No. 502579835
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Theaflavin-3'-Gallate (Theaflavin 2B) usually comes from the leaves of Camellia sinensis (L.) O. Kuntze. Theaflavin-3'-gallate act as prooxidants and induce oxidative stress with carcinoma cells more sensitive than normal fibroblasts. purity: 99%
Catalog No. 50-257-9835 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S05535MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Theaflavin-3'-Gallate (Theaflavin 2B) usually comes from the leaves of Camellia sinensis (L.) O. Kuntze. Theaflavin-3'-gallate act as prooxidants and induce oxidative stress with carcinoma cells more sensitive than normal fibroblasts. purity: 99%