Learn More
Aviva Systems Biology Corp THEG Antibody - middle region (ARP57810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THEG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human THEG. Alternate names: CT56, MGC26138, THEG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-286-389