Learn More
Medchemexpress LLC HY-104066 10mg Medchemexpress, Theliatinib CAS:1353644-70-8 Purity:>98%
SDP
Catalog No. 501968202
Encompass_Preferred
Medchemexpress, HY-104066 10mg Theliatinib CAS:1353644-70-8 Theliatinib (HMPL-309) is a potent, ATP-competitive, orally active and highly selective EGFR inhibitor with a Ki of 0.05 nM and an IC50 of 3 nM. Theliatinib has an IC50 of 22 nM for EGFR T790M/L858R mutant. Theliatinib shows >50-fold selectivity for EGFR than other kinases. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8202 Supplier Medchemexpress LLC Supplier No. HY10406610MG
Medchemexpress, HY-104066 10mg Theliatinib CAS:1353644-70-8 Theliatinib (HMPL-309) is a potent, ATP-competitive, orally active and highly selective EGFR inhibitor with a Ki of 0.05 nM and an IC50 of 3 nM. Theliatinib has an IC50 of 22 nM for EGFR T790M/L858R mutant. Theliatinib shows >50-fold selectivity for EGFR than other kinases. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.