Learn More
Selleck Chemical LLC Theliatinib
SDP
Catalog No. 5000000720
Encompass_Preferred
Theliatinib (HMPL-309) is a highly potent EGFR inhibitor with Ki value of 0 05 nM against the wild type EGFR and IC50 values of 3 nM and 22 nM against EGFR and EGFR T790M/L858R mutant It demonstrats 50 fold greater selectivity for EGFR compared to 72 other kinases
Catalog No. 50-000-00720 Supplier Selleck Chemical LLC Supplier No. S85845MG
Theliatinib (HMPL-309) is a highly potent EGFR inhibitor with Ki value of 0 05 nM against the wild type EGFR and IC50 values of 3 nM and 22 nM against EGFR and EGFR T790M/L858R mutant It demonstrats 50 fold greater selectivity for EGFR compared to 72 other kinases