Learn More
Aviva Systems Biology Corp THEMIS Antibody - C-terminal region (ARP63775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THEMIS. It was validated on Western Blot. . Alternate names: C6orf190, C6orf207, FLJ40584, MGC163388, TSEPA, bA325O24.3, bA325O24.4, GASP, SPOTApplicationWB. Species reactivity: Human
Catalog No. 50-288-080