Learn More
Cole-Parmer Fluke Calibration 1529-R Thermometer, 4 Thermistor/RTD Inputs
Catalog No. 50993455 Shop All Cole-Parmer Products
Encompass
Use this high-accuracy thermometer to read up to four different sensors simultaneously. Display up to eight user-selected data fields such as averages, standard deviations and spreads, probe type and serial number, T1-T2 functions, date, time and battery power levels. Save up to 10 screen configurations for easy recall. Line or battery powered, ideal for both benchtop and on-site readings. Push-button front panel brings up a simple menu that guides you through setup. Choose probe coefficients, sample intervals, communication settings, password protection, and more. RS-232 port allows data download to a computer or printer—order advanced software and printer below. What's included: battery, battery charger, basic software and cable Available for sale in the United States and Canada only.
Catalog No. 50-993-455 Supplier Cole-Parmer Supplier No. 9320210
Add to Cart
Add to Cart
Request a Quote
Use this high-accuracy thermometer to read up to four different sensors simultaneously. Display up to eight user-selected data fields such as averages, standard deviations and spreads, probe type and serial number, T1-T2 functions, date, time and battery power levels. Save up to 10 screen configurations for easy recall. Line or battery powered, ideal for both benchtop and on-site readings. Push-button front panel brings up a simple menu that guides you through setup. Choose probe coefficients, sample intervals, communication settings, password protection, and more. RS-232 port allows data download to a computer or printer—order advanced software and printer below. What's included: battery, battery charger, basic software and cable Available for sale in the United States and Canada only.