Learn More
Selleck Chemical LLC Theobromine S2368-50mg
SDP
Catalog No. 503112713
Encompass_Preferred
Theobromine (3 7-Dimethylxanthine) is a xanthine alkaloid widely consumed as stimulants and snacks in coffee and cocoa based foods and most often as part of ingredients in drugs
Catalog No. 50-311-2713 Supplier Selleck Chemical LLC Supplier No. S236850MG
Theobromine (3 7-Dimethylxanthine) is a xanthine alkaloid widely consumed as stimulants and snacks in coffee and cocoa based foods and most often as part of ingredients in drugs