Learn More
Medchemexpress LLC Theobromine | 83-67-0 | >99.55% | 180.17 | 1 ML
SDP
Catalog No. 5000428566
Encompass_Preferred
Theobromine is a methylxanthine found in cacao beans that can inhibit adenosine receptor A1 (AR1) signaling. It is intended for research use only.
  • Inhibits adenosine receptor A1 (AR1) signaling
  • Decreases lipid accumulation at concentrations above 25 μM without affecting cell viability
  • Suppresses protein expression of PPARγ, C/EBPα, and adipogenic genes at concentrations above 25 μM
  • Lowers body weight and suppresses gains in epididymal and perirenal adipose tissues in vivo
  • Reduces mean adipocyte area in vivo
  • Leads to lower bacterial counts per fecal weight in vivo
  • Increases fecal pH values in vivo
Catalog No. 50-004-28566 Supplier Medchemexpress LLC Supplier No. HYN013810MM/1ML
Theobromine is a methylxanthine found in cacao beans that can inhibit adenosine receptor A1 (AR1) signaling. It is intended for research use only.
  • Inhibits adenosine receptor A1 (AR1) signaling
  • Decreases lipid accumulation at concentrations above 25 μM without affecting cell viability
  • Suppresses protein expression of PPARγ, C/EBPα, and adipogenic genes at concentrations above 25 μM
  • Lowers body weight and suppresses gains in epididymal and perirenal adipose tissues in vivo
  • Reduces mean adipocyte area in vivo
  • Leads to lower bacterial counts per fecal weight in vivo
  • Increases fecal pH values in vivo