Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theobromine, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015651 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5651 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5651 Supplier MilliporeSigma Supelco Supplier No. 42993100MG
Add to Cart
Add to Cart

Theobromine is an active bronchodilator drug ingredient used in the treatment of acute and chronic asthma. It is a naturally present alkaloid in a wide variety of foods and beverages, such as coffee, cocoa, tea, yerba mate and energizing drinks.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 83-67-0
Physical Form Neat
Linear Formula C7H8N4O2
MDL Number MFCD00022830
Melting Point 345°C to 350°C
Synonym 2,6-Dihydroxy-3,7-dimethylpurine; 3,7-Dihydro-3,7-dimethyl-1 H-purine-2,6-dione; 3,7-Dimethylxanthine
Type Analytical Standard
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.