Learn More
Medchemexpress LLC Theodrenaline (hydrochloride) | 2572-61-4 | 411.84 | 25 MG
SDP
Catalog No. 5000449815
Encompass_Preferred
Theodrenaline hydrochloride is a cardiac stimulant that acts as an anti-hypotensive agent when combined with Cafedrine. It induces a positive inotropic effect in human atrial trabeculae by stimulating β-adrenoceptors and can enhance FSK effects, possibly due to PDE-inhibition.
  • Cardiac stimulant
  • Acts as an anti-hypotensive agent when combined with Cafedrine
  • Induces a positive inotropic effect
  • Stimulates β-adrenoceptors
  • Enhances FSK effects, possibly due to PDE-inhibition
  • Appears as a solid
  • For research use only
  • Store at 4°C, protected from light, under nitrogen
Catalog No. 50-004-49815 Supplier Medchemexpress LLC Supplier No. 25MGHYU00344A
Theodrenaline hydrochloride is a cardiac stimulant that acts as an anti-hypotensive agent when combined with Cafedrine. It induces a positive inotropic effect in human atrial trabeculae by stimulating β-adrenoceptors and can enhance FSK effects, possibly due to PDE-inhibition.
  • Cardiac stimulant
  • Acts as an anti-hypotensive agent when combined with Cafedrine
  • Induces a positive inotropic effect
  • Stimulates β-adrenoceptors
  • Enhances FSK effects, possibly due to PDE-inhibition
  • Appears as a solid
  • For research use only
  • Store at 4°C, protected from light, under nitrogen