Learn More
Medchemexpress LLC Theodrenaline hydrochloride | 2572-61-4 | 411.84 | 5 MG
SDP
Catalog No. 5000449149
Encompass_Preferred
Theodrenaline hydrochloride is a cardiac stimulant that also functions as an anti-hypotensive agent when used in conjunction with Cafedrine. It has been shown to evoke a positive inotropic effect in human atrial trabeculae through the stimulation of β-adrenoceptors (AR).
  • Cardiac stimulant
  • Acts as an anti-hypotensive agent when combined with Cafedrine
  • Evokes a positive inotropic effect in human atrial trabeculae by stimulating β-adrenoceptors
  • May potentiate certain effects through phosphodiesterase (PDE) inhibition
  • Targets phosphodiesterase (PDE)
  • Part of the metabolic enzyme/protease pathway
Catalog No. 50-004-49149 Supplier Medchemexpress LLC Supplier No. HYU00344A5MG
Theodrenaline hydrochloride is a cardiac stimulant that also functions as an anti-hypotensive agent when used in conjunction with Cafedrine. It has been shown to evoke a positive inotropic effect in human atrial trabeculae through the stimulation of β-adrenoceptors (AR).
  • Cardiac stimulant
  • Acts as an anti-hypotensive agent when combined with Cafedrine
  • Evokes a positive inotropic effect in human atrial trabeculae by stimulating β-adrenoceptors
  • May potentiate certain effects through phosphodiesterase (PDE) inhibition
  • Targets phosphodiesterase (PDE)
  • Part of the metabolic enzyme/protease pathway