Learn More
Cayman Chemical Theophylline, Purity: ≥98%, CAS: 58-55-9, 25 g
SDP
Catalog No. 502995258
Encompass_Preferred
A phosphodiesterase inhibitor (Ki = 100 µM); an adenosine receptor antagonist (Ki = 14 µM for A1 and A2 receptors); induces relaxation of precontracted feline bronchiole smooth muscle (EC40 = 117 µM; EC80 = 208 µM)
Catalog No. 50-299-5258 Supplier Cayman Chemical Supplier No. 2376025 G
A phosphodiesterase inhibitor (Ki = 100 µM); an adenosine receptor antagonist (Ki = 14 µM for A1 and A2 receptors); induces relaxation of precontracted feline bronchiole smooth muscle (EC40 = 117 µM; EC80 = 208 µM)