Learn More
Medchemexpress LLC Theophylline-d6 | 117490-39-8 | 99.3% | 1 MG
SDP
Catalog No. 5000409471
Encompass_Preferred
Theophylline-d6 is the deuterium labeled Theophylline, a nonselective phosphodiesterase (PDE) inhibitor, adenosine receptor blocker, and histone deacetylase (HDAC) activator. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during the drug development process, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable isotope-labeled compound
  • Used in internal standards for clinical mass spectrometry
  • Deuterium (2H, D) stable isotope-labeled inhibitor
  • Used in therapeutic drug monitoring (TDM)
Catalog No. 50-004-09471 Supplier Medchemexpress LLC Supplier No. HYB0809S1MG
Theophylline-d6 is the deuterium labeled Theophylline, a nonselective phosphodiesterase (PDE) inhibitor, adenosine receptor blocker, and histone deacetylase (HDAC) activator. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during the drug development process, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable isotope-labeled compound
  • Used in internal standards for clinical mass spectrometry
  • Deuterium (2H, D) stable isotope-labeled inhibitor
  • Used in therapeutic drug monitoring (TDM)