Learn More
Medchemexpress LLC Theophylline-d6 | 117490-39-8 | 99.3% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB0809S500UG
Encompass_Preferred
Theophylline-d6 is the deuterium labeled Theophylline. Theophylline is a nonselective phosphodiesterase (PDE) inhibitor, adenosine receptor blocker, and histone deacetylase (HDAC) activator. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08090