Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Theophylline impurity 9 | 42772-85-0 | 216.26 | C9H12O4S | 1 G
SDP

Catalog No. 5000208396
Encompass_Preferred

Theophylline impurity 9 is an analytical standard corresponding to an impurity of the drug theophylline, supplied for research and analytical applications. Consult the Certificate of Analysis for purity and recommended storage conditions.

  • Analytical standard for theophylline impurity 9.
  • Cas number 42772-85-0.
  • Molecular formula C9H12O4S.
  • Molecular weight 216.26.
  • Refer to the certificate of analysis for purity and storage conditions.
  • Packaged quantity: 1 G.

Catalog No. 50-002-08396 Supplier Medchemexpress LLC Supplier No. HYW0874241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Theophylline impurity 9 is an analytical standard corresponding to an impurity of the drug theophylline, supplied for research and analytical applications. Consult the Certificate of Analysis for purity and recommended storage conditions.

  • Analytical standard for theophylline impurity 9.
  • Cas number 42772-85-0.
  • Molecular formula C9H12O4S.
  • Molecular weight 216.26.
  • Refer to the certificate of analysis for purity and storage conditions.
  • Packaged quantity: 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.