Learn More
Medchemexpress LLC Theophylline impurity 9 | 42772-85-0 | >98.0% | 216.26 g/mol | C9H12O4S | 250 MG
SDP
Catalog No. 5000209496
Encompass_Preferred
Theophylline impurity 9 is a characterized impurity of theophylline intended for use in analytical and research applications as a reference or standard. It is provided in small-scale quantities for method development, impurity profiling, and related laboratory work.
  • Used as a reference standard for impurity profiling and analytical testing.
  • Suitable for analytical method development and validation.
  • Available in multiple pack sizes to suit experimental needs.
  • Characterized by molecular formula C9H12O4S and molecular weight 216.26 g/mol.
  • Supplied with a manufacturer datasheet that provides structural and handling information.
Catalog No. 50-002-09496 Supplier Medchemexpress LLC Supplier No. 250MGHYW087424
Theophylline impurity 9 is a characterized impurity of theophylline intended for use in analytical and research applications as a reference or standard. It is provided in small-scale quantities for method development, impurity profiling, and related laboratory work.
  • Used as a reference standard for impurity profiling and analytical testing.
  • Suitable for analytical method development and validation.
  • Available in multiple pack sizes to suit experimental needs.
  • Characterized by molecular formula C9H12O4S and molecular weight 216.26 g/mol.
  • Supplied with a manufacturer datasheet that provides structural and handling information.