Learn More
Medchemexpress LLC Theophylline (Standard) | 58-55-9 | 99.99% | 100 MG
SDP
Catalog No. 5000408117
Encompass_Preferred
Theophylline (Standard) is the analytical standard of Theophylline, intended for research and analytical applications. It functions as a potent phosphodiesterase (PDE) inhibitor, adenosine receptor antagonist, and histone deacetylase (HDAC) activator. It can be used for asthma and chronic obstructive pulmonary disease (COPD) research.
  • Inhibits PDE3 activity to relax airway smooth muscle
  • Possesses anti-inflammatory activity by increasing IL-10 and inhibiting NF-κB into the nucleus
  • Induces apoptosis
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-08117 Supplier Medchemexpress LLC Supplier No. HYB0809R100MG
Theophylline (Standard) is the analytical standard of Theophylline, intended for research and analytical applications. It functions as a potent phosphodiesterase (PDE) inhibitor, adenosine receptor antagonist, and histone deacetylase (HDAC) activator. It can be used for asthma and chronic obstructive pulmonary disease (COPD) research.
  • Inhibits PDE3 activity to relax airway smooth muscle
  • Possesses anti-inflammatory activity by increasing IL-10 and inhibiting NF-κB into the nucleus
  • Induces apoptosis
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS