Learn More
Medchemexpress LLC Theophylline (Standard) | 58-55-9 | 99.99% | 50 MG
SDP
Catalog No. 5000408092
Encompass_Preferred
Theophylline (Standard) is the analytical standard of Theophylline. It is intended for research and analytical applications. It inhibits PDE3 activity to relax airway smooth muscle, has anti-inflammatory activity by increasing IL-10 and inhibiting NF-κB into the nucleus, and induces apoptosis.
  • Potent phosphodiesterase (PDE) inhibitor
  • Adenosine receptor antagonist
  • Histone deacetylase (HDAC) activator
  • Induces apoptosis
  • Used for asthma and chronic obstructive pulmonary disease (COPD) research
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-08092 Supplier Medchemexpress LLC Supplier No. HYB0809R50MG
Theophylline (Standard) is the analytical standard of Theophylline. It is intended for research and analytical applications. It inhibits PDE3 activity to relax airway smooth muscle, has anti-inflammatory activity by increasing IL-10 and inhibiting NF-κB into the nucleus, and induces apoptosis.
  • Potent phosphodiesterase (PDE) inhibitor
  • Adenosine receptor antagonist
  • Histone deacetylase (HDAC) activator
  • Induces apoptosis
  • Used for asthma and chronic obstructive pulmonary disease (COPD) research
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS