Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theophyllol, TCI America™
SDP

Catalog No. T018025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T018025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T018025G Supplier TCI America Supplier No. T018025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8002-89-9
Molecular Formula C9H11N4NaO4
Molecular Weight (g/mol) 262.20
MDL Number MFCD00145398
InChI Key JDYRVPRLEPMNDB-UHFFFAOYSA-M
PubChem CID 91886585
IUPAC Name sodium 1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione acetate
SMILES [Na+].CC([O-])=O.CN1C2=C(NC=N2)C(=O)N(C)C1=O

Specifications

Color White
Quantity 25 g
Formula Weight 284.18
Physical Form Crystalline Powder
Chemical Name or Material Theophyllol
RTECSNumber AJ4550000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.