Learn More
Lonza Walkersville TheraPEAK Lymph Separat Med
Catalog No. 5000496296
Encompass
TheraPEAK Lymphocyte Separation Medium is a GMP produced medium intended for the separation of vital mononuclear cells from whole blood buffy coats or other cell populations Separation is done by low density gradient centrifugation at a density of 1 077 g/ml TheraPEAK Media Products are produced according to applicable GMP raw material standards and are intended to support GMP manufacturing TheraPEAK Media Products are produced at FDA registered manufacturing sites with an ISO 13485 certified quality management system
Catalog No. 50-004-96296 Supplier Lonza Walkersville Supplier No. BP17829E
TheraPEAK Lymphocyte Separation Medium is a GMP produced medium intended for the separation of vital mononuclear cells from whole blood buffy coats or other cell populations Separation is done by low density gradient centrifugation at a density of 1 077 g/ml TheraPEAK Media Products are produced according to applicable GMP raw material standards and are intended to support GMP manufacturing TheraPEAK Media Products are produced at FDA registered manufacturing sites with an ISO 13485 certified quality management system