Learn More
Lonza Walkersville TheraPEAK P3 Primary Cell Nucleofector Solution Set (22.5 mL + 5 mL)
Catalog No. NC2171335
Encompass
TheraPEAK Nucleofector Consumables are aimed to support the use of the 4D-Nucleofector LV Unit as manufacturing equipment in a GMP manufacturing process for advanced therapeutic medicinal products (ATMPs).In such a process, the 4D-Nucleofector LV Unit and its consumables may be used for non-viral, electroporation-based genetic modification of primary cells, for example T cells or hematopoietic stem cells.Benefits:Chemically-defined, human and animal-component free compositionIdentity tested, Ph.Eur. grade raw materialsAseptically produced at a GMP certified site in a class A environmentFilled in crimp-sealed glass vialsDouble-layer packaging supporting the clean room cascadeValidated manufacturing and testing methodsApplications:Supporting use of 4D-Nucleofector LV Unit in GMP manufacturingContent:22.5 mL TheraPEAK P3 Nucleofector Solution5 mL TheraPEAK Nucleofector Supplement 1
This item is not returnable. View return policy
Catalog No. NC2171335 Supplier Lonza Walkersville Supplier No. G4LP322500
TheraPEAK Nucleofector Consumables are aimed to support the use of the 4D-Nucleofector LV Unit as manufacturing equipment in a GMP manufacturing process for advanced therapeutic medicinal products (ATMPs).In such a process, the 4D-Nucleofector LV Unit and its consumables may be used for non-viral, electroporation-based genetic modification of primary cells, for example T cells or hematopoietic stem cells.Benefits:Chemically-defined, human and animal-component free compositionIdentity tested, Ph.Eur. grade raw materialsAseptically produced at a GMP certified site in a class A environmentFilled in crimp-sealed glass vialsDouble-layer packaging supporting the clean room cascadeValidated manufacturing and testing methodsApplications:Supporting use of 4D-Nucleofector LV Unit in GMP manufacturingContent:22.5 mL TheraPEAK P3 Nucleofector Solution5 mL TheraPEAK Nucleofector Supplement 1