Learn More
Lonza Walkersville TheraPEAK T-VIVO Cell Culture Medium, chemically defined, serum-free, non-animal origin, 1 L bottle
Supplier:  Lonza Walkersville BP12970Q
Encompass
The Lonza TheraPEAK T-VIVO Cell Culture Medium is a specialized formulation designed for optimal T-cell culture and expansion in cell therapy applications. This medium is chemically defined, serum-free, and derived from non-animal sources, ensuring enhanced cell proliferation and viability for various hematopoietic cells, including T cells and CAR-T cells. Its formulation facilitates compliance with regulatory standards and simplifies scale-up from research to manufacturing.
  • Chemically defined formulation
  • Serum-free, non-animal origin
  • Supports T cells, CAR-T cells, PBL, TIL growth
  • High transduction efficiencies
  • Produced under GMP guidelines
  • ISO 13485 certified quality management
  • Compatibility with various culture platforms
  • Storage at 2-8 °C, protected from light
  • Available in a 1 L bottle
  • Not for human or animal in vivo use
Catalog No. NC3308140